Merck
Z229695
Sigma-Aldrich
Aldrich® hollow pennyhead stopper
joint: ST/NS 29/32
Synonym(e):
glass stopper, glass stoppers, stoppers
About This Item
UNSPSC-Code:
41122500
NACRES:
NB.35
Materialien
ground glass
Verbindung
joint: ST/NS29/32
Beschreibung
closed at bottom
Allgemeine Beschreibung
Conforms to standard-taper specifications as outlined in ASTM E 675.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)