Merck
Z183601
Sigma-Aldrich
Cryo-apron®
size 24 in. × 42 in.
Synonym(e):
Cryogenic Apron
About This Item
UNSPSC-Code:
46181501
NACRES:
NB.35
Verpackung
pack of 1 ea
Hersteller/Markenname
TempShield CA42
Größe
24 in. × 42 in.
Allgemeine Beschreibung
Provides protection in extreme environments. Resistant to solvents/chemicals. Breathable, static-free, and asbestos-free. 100% waterproof. Reversible. Adjustable straps at neck and waist.
Rechtliche Hinweise
Cryo-apron is a registered trademark of Tempshield, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)