Y906514
Valve
High Pressure, brass diaphragm, 1/4 in. NPT straight path, external
Alle Fotos(1)
About This Item
UNSPSC-Code:
12142200
NACRES:
NA.12
Empfohlene Produkte
Materialien
brass diaphragm
Qualitätsniveau
Parameter
3000 psi max. pressure
Länge
2.387 in.
Gewicht
225 g
Fitting
1/4 in. NPT straight path, external
Verwandte Kategorien
Sonstige Hinweise
This product is only available with the purchase of a bulk gas product and not for individual sale. For more information, please contact Stable Isotopes Customer Service
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)