Merck
WHA10461000
Whatman® FP 025/1 filter holder
Synonym(e):
Z698504
About This Item
UNSPSC-Code:
41104922
NACRES:
NB.24
Sterilität
non-sterile
Verpackung
pkg of 10 ea
Hersteller/Markenname
Whatman 10461000
Whatman Article No. 28415718 (US reference)
Allgemeine Beschreibung
The syringe filter holder has a female Luer lock inlet and male Luer outlet compatible with typical syringes. Polysulfone housing can be autoclaved with membrane in place for applications requiring a sterile filter assembly.
Leistungsmerkmale und Vorteile
  • Durable polysulfone plastic construction provides moderate chemical compatibility with lighter weight than stainless steel filter holders.
  • Polysulfone filter holders are often used for clarification, venting applications, particulate removal or collection from aqueous, air or gas streams.
Sonstige Hinweise
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Rechtliche Hinweise
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Lot/Batch Number
Leider sind derzeit keine COAs für dieses Produkt online verfügbar.
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
Slide 1 of 1