Merck
TEM-FF100-NI-50
Sigma-Aldrich
Formvar Supported Nickel Square Mesh
size 100 mesh, box of 50
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Verpackung
box of 50
Größe
100 mesh
Allgemeine Beschreibung
A 10 nm thin formvar film supported standard square mesh grids. Nickel grid, 100 mesh. All of our film is laid on the shiny side of the grid.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)