SYNV381512
Synthware™ vacuum trap with #15 o-ring joint and high vacuum valve
body O.D. 57 mm, body L 300 mm
Synonym(e):
Synthware V381512
Alle Fotos(1)
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.51
Empfohlene Produkte
Hersteller/Markenname
V381512
Grundkörper L
300 mm
Grundkörper AD
57 mm
Verwandte Kategorien
Allgemeine Beschreibung
Synthware™ vacuum trap features a two-piece design with a 0-12 mm high vacuum valve as the shut-off. Connection is made via a 57 mm O-ring flat flange and a #331 Viton® O-ring.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)