SYNV163229
Synthware™ vacuum trap with serrated hose connections
joint: ST/NS 29/42, body O.D. 32 mm, L 200 mm , below joint
Synonym(e):
Synthware V163229
Alle Fotos(1)
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.51
Empfohlene Produkte
Verbindung
joint: ST/NS29/42
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware V163229
Länge
200 mm , below joint
Grundkörper AD
32 mm
Verwandte Kategorien
Allgemeine Beschreibung
With joints for easy removal of condensate. Serrated hose connections for side tube have an O.D. of 13 mm at the smallest serration and 16 mm at the largest serration. Center tube hose connection is always 10mm for easy connection to manifold bank tubes which are also with 10 mm hose connection.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)