SYNV150500
Synthware™ photochemical reaction vessel with internal thread
500 mL
Synonym(e):
Synthware™ Double-Walled Quartz Photochemical Immersion Well,, Synthware V150500
Alle Fotos(1)
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.51
Empfohlene Produkte
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware V150500
Kapazität
500 mL
Allgemeine Beschreibung
Reaction Vessel constructed of borosilicate glass to accommodate immersion well. With #50 internal thread at top which accepts a RC50 bushing and Viton® O-ring to form a compression seal with the immersion well, one 14/20 joint for sparger tube, one 24/40 vertical joint for condenser and one #7 internal thread side arm for thermometer. Complete with quartz immersion well, caps and O-rings.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
SYNV150500-1EA | 4061832209029 |