SYNM190042
Synthware™ vacuum manifold with PTFE stopcocks
number of ports, 4, Hose Connection: right, stopcock size 2 mm
Synonym(e):
Synthware M190042, vacuum manifold, vacuum unit
Alle Fotos(1)
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.51
Empfohlene Produkte
Leistungsmerkmale
number of ports 4
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware M190042
Absperrhahngröße
2 mm
Verwandte Kategorien
Allgemeine Beschreibung
General purpose manifold with 2 mm or 4 mm PTFE stopcocks. Main tube is 22 mm O.D. with serrated hose connection(10 mm O.D.) at right side. Delivery ports have 10 mm serrated hose connections. Distance between stopcocks is 75mm.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
SYNM190042-1EA | 4061832196992 |