Merck
SYNA852440
Synthware U-shaped drying tube
joint: ST/NS 24/40
Synonym(e):
Synthware A852440, Drying tube adapter
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Materialien
colorless tube
glass tube
Verbindung
joint: ST/NS24/40
Sterilität
non-sterile
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A852440
Allgemeine Beschreibung
With joint. Cotton is usually used to contain drying agent.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)