MT51344295
Mettler-Toledo electrode cable
connector AK9, BNC-RCA
Alle Fotos(1)
About This Item
UNSPSC-Code:
41115600
NACRES:
NB.61
Empfohlene Produkte
Hersteller/Markenname
Mettler-Toledo 51344295
Allgemeine Beschreibung
Mettler Toledo electrodes can easily be connected to various third-party instruments. All you have to do is select the appropriate cable. We offer a wide choice of cable lengths and plug combinations. For Mettler Toledo reference electrodes and temperature probes.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)