Merck
MT51343010
Mettler Toledo InLab® Easy pH electrode
S7 connector, without cable
About This Item
UNSPSC-Code:
41115604
NACRES:
NB.79
Materialien
glass, U membrane
polysulfone shaft
Hersteller/Markenname
Mettler-Toledo 51343010
Temperaturmessbereich
0-80 °C
L × B
120 mm × 12 mm
pH-Wert
0-14
Allgemeine Beschreibung
The Mettler Toledo InLab® Easy are low maintenance, pH combination electrodes with polysulphone shaft, gel electrolyte, ceramic junction and variety of connectors. Ideal for field measurement and educational use
Ideal for field measurement and educational use.
The tough, durable polysulphone shaft with integral bulb protector gives maximum protection against breakage. The use of a gel electrolyte avoids the inconvenience of re-filling the reference electrolyte in the field. Different cables can be connected to the S7 screw head allowing use with a wide range of meters.
  • Robust polysulphone shaft
  • Low maintenance gel electrolyte
  • \Integral bulb protector
  • Detachable cable
Rechtliche Hinweise
InLab is a registered trademark of Mettler-Toledo, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)