Merck
GF96869424
Quartz - fused
sheet, thickness 3.0 mm, size 150 × 150 mm, condition polished both sides
Synonym(e):
SI613141
About This Item
UNSPSC-Code:
11101503
NACRES:
NA.23
Form
sheet
Hersteller/Markenname
Goodfellow 968-694-24
Größe
150 × 150 mm
Dicke
3.0 mm