Merck
DWK227494-03G
Wheaton® Bod Bottle
capacity 60 mL, bottle diam. × H 43 mm × 115 mm, pennyhead glass cap (stopper)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
pennyhead glass cap (stopper)
Agentur
suitable for SM 5210
Verpackung
pkg of 36 ea
Hersteller/Markenname
WHEATON® 227494-03G
Flaschendurchm. × H
43 mm × 115 mm
Kapazität
60 mL
Allgemeine Beschreibung
BOD Bottles are ideal for incubating diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during the stabilization of the decomposable organic matter by aerobic biochemical action. We recommend the 300mL capacity bottles for the five-day BOD test as referenced in Standard Methods for the Examination of Water and Wastewater, 21st Edition, 5210B, 2005.
Leistungsmerkmale und Vorteile
Ideal for incubating diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during the stabilization of the decomposable organic matter by aerobic biochemical action
Rechtliche Hinweise
WHEATON is a registered trademark of Wheaton Industries, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)