CLS72502L
Pyrex® Mallory direct reading settlometer
volume 2000 mL
Synonym(e):
lab equipment accessory
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Empfohlene Produkte
Hersteller/Markenname
Corning 7250-2L
Volumen
2000 mL
Verwandte Kategorien
Allgemeine Beschreibung
Used with oxidized sludge process for treatment of sewage and other wastes. 2000 mL heavy wall cylinder measures 51/8". I.D. × 71/2". high and is graduated in cc per liter, and in hundredths of a foot.
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)