Merck
CLS6383250
Pyrex® cylindrical separatory funnel, PTFE stopcock
Synonym(e):
Corning® 6383
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.43
Verbindung
joint stopper22
Hersteller/Markenname
Corning 6383-250
Kapazität
250 mL
Absperrhahngröße
2
Allgemeine Beschreibung
Graduated, with ST Pyrex stopper and microfinish PTFE stopcock.
CLS6383A100 - does not include a stopper
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)