Merck
CLS409000
Corning® PTFE-coated magnetic stir bar
L × O.D. 3 in. × 3/4 in.
Synonym(e):
Corning stirrer accessory
About This Item
UNSPSC-Code:
41122401
NACRES:
NB.72
Materialien
PTFE-coated
oval
Hersteller/Markenname
Corning 409000
L × AD
3 in. × 3/4 in.
Allgemeine Beschreibung
These 2 x 3/8 inch (5.08 x 0.95 cm) Corning magnetic stir bars are Polytetrafluorethylene (PTFE)-coated for increased chemical resistance.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
CLS409000-1EA4061825881294