Merck
CLS401661
Corning® Modified Impeller Assembly (for use with probes) for Magnetically-Driven Proculture® Spinner Flask
fits flask, 15 L
Synonym(e):
equipment replacement accessory
About This Item
UNSPSC-Code:
41122104
NACRES:
NB.43
Materialien
stainless steel shaft
Sterilität
non-sterile
Leistungsmerkmale
fits flask 15 L
Hersteller/Markenname
Corning 401661
Allgemeine Beschreibung
Stainless steel impeller shaft with dual bearing, modified impeller assembly for use in magnetically driven, 15L Proculture Spinner Flasks. Narrower impeller paddle design allows use with probes to create a small bioreactor.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)