Merck
BR722055
BRAND® sample vial
for Coulter-Counter, volume 20 mL, with lid
Synonym(e):
Sample Cups
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.11
Materialien
polystyrene
Beschreibung
4 bags of 250 pack quantity 1000 pcs
Sterilität
non-sterile
Verpackung
pack of 1000 ea
Hersteller/Markenname
BRAND 722055
Volumen
20 mL
Allgemeine Beschreibung
The BRAND® Sample Vial is designed for secure and efficient storage of samples in laboratory environments. Made from high-quality transparent polystyrene (PS), this vial allows for easy visibility of contents, facilitating quick identification and monitoring. It features a polyethylene (PE) lid that provides a secure closure, ensuring sample integrity and preventing contamination.
Leistungsmerkmale und Vorteile
  • BRAND® sample vial is designed for various laboratory applications.
  • Made of transparent polystyrene for accurate and quick sample analysis.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)