Merck
BR719805
BRAND® counting chamber BLAUBRAND® Fuchs-Rosenthal
without clips, double ruled
Synonym(e):
BLAUBRAND®, Cell culture, Counting Chambers, Fuchs-Rosenthal
About This Item
UNSPSC-Code:
41122600
NACRES:
NB.22
Materialien
colorless
glass
Sterilität
non-sterile
Leistungsmerkmale
without clips
Hersteller/Markenname
BRAND 719805
Wells
1
Allgemeine Beschreibung
The ruling differs from the customary systems for blood cell counts by its large area of 16 mm2.The ruling shows 16 large squares of 1 mm2. Each large square is subdivided into 16 mini squares with 0.25 mm sides and an area of 0.0625 mm2.This chamber is frequently used for counting cells in the cerebro-spinal fluid.
Leistungsmerkmale und Vorteile
  • BLAUBRAND® quality
  • Outstanding measurement precision
  • 100% certified quality
Verpackung
1 counting chamber, complete with 2 haemacytometer cover glasses, in transparent plastic box.
Rechtliche Hinweise
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)