Merck
BR704551
BRAND® replacement pump assembly for seripettor® pro
size 2 mL
Synonym(e):
Accessories, Bottle-top Dispenser, Dispenser, seripettor®
About This Item
UNSPSC-Code:
40151500
NACRES:
NB.43
Materialien
(Hastalloy (stainless))
(Polyphenylene Oxide / Polyetherimide (UV protection))
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 704551
Größe
2 mL
Verwandte Kategorien
Allgemeine Beschreibung
The BRAND® Replacement Pump Assembly for the Seripettor® Pro is designed for precise and efficient liquid handling in laboratory applications. This actuating unit is constructed from high-quality materials, including polyphenylene oxide (PPO) and polyetherimide (PEI) for UV protection, ensuring durability and longevity. The assembly features a Hastelloy® lifting spring, which enhances performance and reliability. This replacement pump assembly is essential for maintaining the functionality of your Seripettor® Pro, making it an indispensable tool for researchers and technicians seeking accurate and efficient liquid dispensing.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hastelloy is a registered trademark of Haynes International, Inc.
seripettor is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)