Merck
BR701928
BRAND® replacement piston rod for TRANSFERPETTOR
for fixed type 1, 2, 3, 4, 5 μL
Synonym(e):
Accessories, Pipettes, Piston rod, Piston-operated pipettes, Positive displacement principle, Transferpettor
About This Item
UNSPSC-Code:
41121501
NACRES:
NB.01
Materialien
stainless steel
Verpackung
pack of 3 ea
Hersteller/Markenname
BRAND 701928
Allgemeine Beschreibung
The BRAND® Replacement Piston Rod for the TRANSFERPETTOR is designed to ensure optimal performance and precision in liquid handling. Starting from 20 µl, product comes equipped with a seal. The replacement rod is easy to install and maintain, providing a reliable solution for users seeking to extend the life of their TRANSFERPETTOR while maintaining accuracy and efficiency in their pipetting tasks.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)