BR470050
BRAND® cover glass
L × W 20 mm × 20 mm, square
Synonym(e):
Cover glasses, Microscopy
Alle Fotos(1)
About This Item
UNSPSC-Code:
41122600
NACRES:
MA.01
Empfohlene Produkte
Materialien
borosilicate glass
square
Hersteller/Markenname
BRAND 470050
L × B
20 mm × 20 mm
Dicke
0.13-0.17 mm , No. 1
Verwandte Kategorien
Allgemeine Beschreibung
Clear borosilicate glass, hydrolytic class 1, with excellent chemical resistance. Thickness No. 1 (0.13 to 0.17 mm). Refractive index ne = 1.52 ± 0.01, Abbe number ve = 56.5 ± 0.5. Full automatic production guarantees "ready-for-use" quality - clean, dust and grease free, distortion-free, flatness within ± 3 µm.
Leistungsmerkmale und Vorteile
- BRAND® borosilicate cover glass offers superior clarity and excellent chemical resistance.
- Cover glasses for microscope slides are clean, dust/grease free and distortion-free.
- Full automatic production guarantees "ready-for-use" quality.
- Flatness within ± 3 μm.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
BR470050-2000EA | 4061831857115 |