BR44870
BRAND® Pursept™-A disinfectant
XL towel dispenser refill pack
Synonym(e):
Pursept® Wipes XL
Alle Fotos(1)
About This Item
UNSPSC-Code:
47131800
NACRES:
NB.35
Empfohlene Produkte
Verpackung
pack of 70 ea
Hersteller/Markenname
BRAND 44870
Größe
230 mm × 280 mm
Allgemeine Beschreibung
Fleece wipes for impregnation with universal or disinfecting detergents.The lint-free and tear-proof wipes are designed for both optimal disinfectant delivery and excellent contaminant uptake. Due to their size they are particularly suitable to disinfect your workplace, lab table or clean bench quickly and reliably.The wipes can be stored in the convenient dispenser box.For high-volume use, wipes can be soaked with liquid disinfectant directly in the handy dispenser box (lasts up to 28 days).We recommend the usage of the surface disinfecting detergent Pursept® A Xpress.Technical Information:Fleece wipes made of 100% polyester, dry.70 wipes (230 x 280 mm) per roll.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Pursept is a registered trademark of Merz GmbH & Co. KG
p3xFLAG is a trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR44870-6ROLL | 4061831856576 |