Merck
BR365538
BRAND® volumetric flask for oil content determination
capacity 100 mL
Synonym(e):
Flasks, SILBERBRAND, Volumetric flasks
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
glass
Verbindung
joint ST/NS neck19/26
Leistungsmerkmale
accuracy class B
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 365538
Kapazität
100 mL
Allgemeine Beschreibung
Borosilicate glass 3.3. To DIN 51368. Interchangeable hollow glass stopper size 19/26. Calibrated to contain (TC, In). For determination of oil content in aqueous oil emulsions (e.g. drilling oil). EL = Error limits.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)