Merck
BR130264
BRAND® amber narrow-mouth bottle, LDPE, with screw cap
capacity 2000 mL
Synonym(e):
Bottles, Plastics
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
Materialien
low-density polyethylene cap
round amber low-density polyethylene bottle
Verbindung
Joints threaded neck (GL28)
Leistungsmerkmale
with cap
Verpackung
pack of 100 ea
Hersteller/Markenname
BRAND 130264
Kapazität
2000 mL
Durchm. × H
117 mm × 264 mm
Allgemeine Beschreibung
For storage of light-sensitive substances. With screw cap
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)