Merck
BR129273
BRAND® screw cap wash head for wash bottles of PFA
GL 25
Synonym(e):
Accessories, Bottles, Screw caps
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
Materialien
ETFE screw cap
FEP tube
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 129273
Fitting
GL 25
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)