Merck
BAH500250409
SP Bel-Art® Glove Box Sleeved Gloves for 8 in. Glove Ports
size 9 medium
Synonym(e):
glove box
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.43
Materialien
black neoprene
Größe
9 medium
Verpackung
pkg of 1 pair
Hersteller/Markenname
Bel-Art H50025-0409
Allgemeine Beschreibung
One-piece, seamless, smooth neoprene protection against liquid, gas or solid substance penetration.
Leistungsmerkmale und Vorteile
  • Suitable for use with glove boxes having 8" glove ports including sid ENTRY H50026-0000 Glove Boxes
  • Requires clamping rings (H50025-0320) to hold gloves in place
  • Sold in pairs
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)