Merck
BAH367140000
SP Bel-Art® Quaver Vibrating Spatula
stainless steel blade
Synonym(e):
spoon
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.43
Materialien
stainless steel blade
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art H36714-0000
Allgemeine Beschreibung
Make accurate additions of precious materials as needed. Rotating the toothed thumb wheel vibrates the curved tip spatula, giving you control over transfer from spatula to weighing dish.
Leistungsmerkmale und Vorteile
  • Available with a plain stainless steel blade or with a brown FEP coating
  • Coating complies with FDA regulations governing direct food contact applications; reduces transfer losses
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)