Merck
BAF369740600
SP Bel-Art® Sterileware Powder Lance Samplers
white high-density polyethylene, capacity 125 mL (4 oz), L 23.625 in. (60 cm), sterile, pack of 20 ea (individually wrapped)
Synonym(e):
sampler
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.35
Materialien
white high-density polyethylene
Sterilität
sterile
Verpackung
pack of 20 ea (individually wrapped)
Hersteller/Markenname
Bel-Art F36974-0600
Länge
23.625 in. (60 cm)
Kapazität
125 mL (4 oz)
Allgemeine Beschreibung
A simple insert, rotate, and withdraw motion allows cross sectional sampling of non-flowing, bulk powders. Available in three lengths.
Leistungsmerkmale und Vorteile
  • Open blade works well with sticky, cohesive powders
  • Rounded edges help hold sample in place
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) lance is heat sealed in individual polyethylene bag
  • Gamma irradiated sterile (SAL 10^-6)
  • Ready for one time use and disposal
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)