Merck
BAF116340000
Spray pump bottles
capacity 500 mL
Synonym(e):
bottle pump, spray bottle
About This Item
UNSPSC-Code:
41103209
NACRES:
NB.17
Leistungsmerkmale
with cap
Verpackung
pack of 12 ea
Hersteller/Markenname
Bel-Art F11634-0000
Kapazität
500 mL
Allgemeine Beschreibung
To use this low-density plunger polyethylene (LDPE) bottle, simply press the plunger to dispense a misty spray. Clear cap helps prevent evaporation and accidental leaks.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)