ACE6893021
Ace receiving flask for rotary evaporators, graduated, round bottom
size 1000 mL
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Empfohlene Produkte
Materialien
borosilicate glass flask
round bottom
single-neck
Verbindung
joint 35/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Ace Glass 6893-21
Größe
1000 mL
Verwandte Kategorien
Allgemeine Beschreibung
Graduated, round bottom receiving flask for use with all rotary evaporators. 35/20 spherical outer joint will also mate to 35/25 ball joints. 250 mL and 500 mL size graduated in 10 mL increments, 1000 mL and 2000 mL in 100 mL increments.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
ACE6893021-1EA | 4061832346151 |