ACE585306
Ace-Safe® tubing connector, UHDPE
#7 to 1/4 in. I.D. tubing
Alle Fotos(1)
About This Item
UNSPSC-Code:
41106514
NACRES:
NB.79
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
Ace Glass 5853-06
Parameter
-20-110 °C temp. limit
Verwandte Kategorien
Allgemeine Beschreibung
Tubing connector, used to connect flexible tubing (1/4 in., 3/8 in., 1/2 in., 3/4 in., 1 in. I.D.) to #7, #11, #15 or #25 Ace-Thred for easy, safe connect/disconnect. Nylon bushing slides over serrated end and secures PP connector in thread with silicone O-ring in front groove to make compression seal. With tubing attached at serrated end, un-thread bushing and connector is removed from thread.
Rechtliche Hinweise
Ace-Safe is a registered trademark of Ace Glass, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)