Skip to Content
Merck
All Photos(1)

Key Documents

713-0089

GC Analysis of Menthol Enantiomers on α-DEX 120

suitable for GC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

gas chromatography (GC): suitable

test parameters

column: α-DEX 120, 30 m × 0.25 mm I.D., 0.25 μm (24310)
oven: 110 °C
detector: FID, 300 °C
carrier gas: helium, 20 cm/sec
injection: 1 μL, 100:1 split
sample: 0.5 mg/mL each component

application(s)

cleaning products
cosmetics
flavors and fragrances
personal care

Legal Information

DEX is a trademark of Sigma-Aldrich Co. LLC

Analyte

Description

    (+)-Neomenthol
    (−)-Menthol
    (−)-Neomenthol
    (+)-Menthol

analytical column

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service