Skip to Content
Merck
All Photos(8)

Key Documents

ZRB2969

Sigma-Aldrich

Anti-PDX1 Antibody, clone 1I16 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

GSF, Glucose-sensitive factor, IDX-1, IPF-1, IUF-1, Insulin promoter factor 1, Insulin upstream factor 1, Islet/duodenum homeobox-1, Pancreas/duodenum homeobox protein 1, STF-1, Somatostatin-transactivating factor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1I16, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 30.77 kDa
observed mol wt ~38 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey, bovine, porcine

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... PDX1(3651)

Related Categories

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1I16 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Pancreas/duodenum homeobox protein 1 (PDX1). It targets an epitope within 17 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the N-terminal region of human Pancreas/duodenum homeobox protein 1 (PDX1).

Application

Quality Control Testing

Evaluated by Western Blotting in Caco-2 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PDX1 in Caco-2 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PDX1 in PANC-1 cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PDX1 in PANC-1 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected PDX1 in human pancreas tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound PDX1 peptide with a KD of 2.9 x 10-9 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Pancreas/duodenum homeobox protein 1 (UniProt: P52945; also known as PDX-1, Glucose-sensitive factor, GSF, Insulin promoter factor 1, IPF-1, Insulin upstream factor 1, IUF-1, Islet/duodenum homeobox-1, IDX-1, Somatostatin-transactivating factor 1, STF-1) is encoded by the PDX1 (also known as IPF1, STF1) gene (Gene ID: 3651) in human. PDX1 is a transcription factor that serves as a master regulator of pancreas organogenesis, maturation, and identity preservation of β-cells. It is involved in activation of insulin, somatostatin, glucokinase, and GLUT2 gene transcription. Although it is mainly expressed in β-cells and some -cells of the islets of Langerhans, its expression is also observed in duodenum. It is expressed early in pancreas development and persists through β-cell maturation. It is detected both in the nucleus and cytoplasm. Its nuclear localization signal resides in amino acids 197-203 and its homeobox domain is localized in amino acids 146-205. Glucose induces its nuclear translocation by triggering its phosphorylation via the PI3-kinase pathway. It is reported to be essential for compensatory β cell mass expansion in response to diet-induced insulin resistance through its roles in promoting β cell survival and compensatory hypertrophy. Mutations in PDX1 gene are linked to pancreatic agenesis that can lead to early-onset insulin-dependent diabetes mellitus (IDDM), as well as maturity onset diabetes of the young type 4 (MODY4). PDX1 deficient β cells show evidence of endoplasmic reticulum (ER) stress. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Ebrahim, N., et al. (2022). Front. Mol. Biosci. 9;1091757; Sachdeva, MM., et al. (2009). Proc. Natl. Acad. Sci. USA. 106(45); 19090-19095).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service