Z330094
BRAND® wide-mouth bottles, LDPE
capacity 100 mL, with cap
Synonym(s):
ldpe bottle, polyethylene bottles, polyethylene containers
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
low-density polyethylene bottle
round bottle
translucent bottle
wide-mouth bottle
Joint
Joints threaded neck (GL32)
feature
with cap
packaging
pkg of 50 ea
manufacturer/tradename
BRAND 129638
capacity
100 mL
diam. × H
48 mm × 94 mm
color
natural
Looking for similar products? Visit Product Comparison Guide
General description
Flexible, translucent.
Other Notes
BR129638 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service