Skip to Content
Merck
All Photos(2)

Key Documents

X0879

Sigma-Aldrich

Anti-XPB antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-BTF2, Anti-ERCC3, Anti-Excision repair cross-complementing rodent repair deficiency, complementation group 3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 90 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

indirect immunofluorescence: 10-20 μg/mL using HeLa cells fixed with paraformaldehyde-Triton
western blot: 1-2 μg/mL using HepG2 cell lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ERCC3(2071)
mouse ... Ercc3(13872)
rat ... Ercc3(291703)

General description

XPB (Xeroderma pigmentosum complementation group B) is located on the human chromosome at 2q14.3. TFIIH (General transcription and DNA repair factor IIH helicase) consists of nine protein subunits, XPB (ERCC3) being one of them. It is an ATP-dependent DNA helicase of 89 kDa. The XPB is localized in the neurons.

Immunogen

synthetic peptide corresponding to amino acids 765-782 of human XPB, also known as ERCC3, conjugated to KLH via an N-terminal added cysteine residue. The immunizing peptide differs from the mouse and rat sequences in one and three amino acids respectively.

Application

Anti-XPB antibody has been used in immunoblotting andimmunofluorescence.

Biochem/physiol Actions

XPB (General transcription and DNA repair factor IIH helicase subunit XPB) is involved in DNA repair and transcription. Phosphorylation on Ser751 of XPB inhibits TFIIH repair activity, leaving its transcription function intact. Mutations in the gene encoding XPB causes Xeroderma Pigmentosum, Cockayne′s syndrome, and trichothiodystrophy.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DNA-based diagnosis of xeroderma pigmentosum group C by whole-genome scan using single-nucleotide polymorphism microarray
Lam C W, et al.
The Journal of Investigative Dermatology, 124(1), 87-91 (2005)
Immunohistochemical analysis of nucleotide excision repair factors XPA and XPB in adult rat brain
Yang S Z, et al.
Anatomical Record (Hoboken, N.J. : 2007), 291(7), 775-780 (2008)
Structural Basis for S100B Interaction with its Target Proteins
Prez K D and Fan L
Journal of Molecular and Genetic Medicine, 12(3), 775-780 (2018)
Luisa Mori et al.
Journal of virology (2020-11-27)
HIV transcription requires assembly of cellular transcription factors at the HIV-1promoter. The TFIIH general transcription factor facilitates transcription initiation by opening the DNA strands around the transcription start site and phosphorylating the C-terminal domain for RNA polymerase II (RNAPII) for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service