Skip to Content
Merck
All Photos(5)

Key Documents

SAB4200518

Sigma-Aldrich

Anti-SLC1A1 antibody produced in rabbit

enhanced validation

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-EAAT3, Anti-solute carrier family 1, member 1, EAAC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
WB
clone:
polyclonal
species reactivity:
mouse, human
citations:
2
technique(s):
immunohistochemistry: 10-20 μg/mL using formalin-fixed paraffin-embedded human small intestine.
western blot: 1.5-3.0 μg/mL using Neuro-2a cell extracts.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~70 kDa

species reactivity

mouse, human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: 10-20 μg/mL using formalin-fixed paraffin-embedded human small intestine.
western blot: 1.5-3.0 μg/mL using Neuro-2a cell extracts.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC1A1(6505)
mouse ... Slc1a1(20510)
rat ... Slc1a1(25550)

General description

SLC1A1 (also known as EAAC1, EAAT3) is localized at the postsynaptic site of neurons. It is expressed at moderate levels in the adult brain, mainly in the hippocampus, basal ganglia and olfactory bulb and is considered to make a minor contribution to glutamate removal from the synapse.

Immunogen

synthetic peptide corresponding to a sequence near the C-terminus of human SLC1A1, conjugated to KLH. The corresponding sequence is identical in mouse and rat SLC1A1.

Application

Anti-SLC1A1 antibody produced in rabbit has been used in immunohistochemistry and immunoblotting.

Biochem/physiol Actions

SLC1A1 regulates the developmental effects of glutamate. It is highly regulated by neural activity and intracellular signaling pathways involving mainly PKCa, PI3K, and glial factors, resulting in rapid changes in the trafficking of the transporter and in its membrane location.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Plasma membrane expression of the neuronal glutamate transporter EAAC1 is regulated by glial factors: evidence for different regulatory pathways associated with neuronal maturation
Lortet S, et al.
Neurochemistry International, 52(7), 1373-1382 (2008)
The neuronal excitatory amino acid transporter EAAC1/EAAT3: does it represent a major actor at the brain excitatory synapse?
Nieoullon A, et al.
Journal of Neurochemistry, 98(4), 1007-1018 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service