Skip to Content
Merck
All Photos(1)

Key Documents

SAB1405546

Sigma-Aldrich

Anti-CACNB4 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CAB4, CACNLB4, EA5, EJM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~58.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CACNB4(785)

General description

This gene encodes a member of the beta subunit family of voltage-dependent calcium channel complex proteins. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Various versions of each of these subunits exist, either expressed from similar genes or the result of alternative splicing. The protein encoded by this locus plays an important role in calcium channel function by modulating G protein inhibition, increasing peak calcium current, controlling the alpha-1 subunit membrane targeting and shifting the voltage dependence of activation and inactivation. Certain mutations in this gene have been associated with idiopathic generalized epilepsy (IGE) and juvenile myoclonic epilepsy (JME). Multiple transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

CACNB4 (NP_000717.2, 1 a.a. ~ 520 a.a) full-length human protein.

Sequence
MSSSSYAKNGTADGPHSPTSQVARGTTTRRSRLKRSDGSTTSTSFILRQGSADSYTSRPSDSDVSLEEDREAIRQEREQQAAIQLERAKSKPVAFAVKTNVSYCGALDEDVPVPSTAISFDAKDFLHIKEKYNNDWWIGRLVKEGCEIGFIPSPLRLENIRIQQEQKRGRFHGGKSSGNSSSSLGEMVSGTFRATPTSTAKQKQKVTEHIPPYDVVPSMRPVVLVGPSLKGYEVTDMMQKALFDFLKHRFDGRISITRVTADISLAKRSVLNNPSKRAIIERSNTRSSLAEVQSEIERIFELARSLQLVVLDADTINHPAQLIKTSLAPIIVHVKVSSPKVLQRLIKSRGKSQSKHLNVQLVAADKLAQCPPEMFDVILDENQLEDACEHLGEYLEAYWRATHTTSSTPMTPLLGRNLGSTALSPYPTAISGLQSQRMRHSNHSTENSPIERRSLMTSDENYHNERARKSRNRLSSSSQHSRDHYPLVEEDYPDSYQDTYKPHRNRGSPGGYSHDSRHRL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service