Skip to Content
Merck
All Photos(2)

Key Documents

HPA026293

Sigma-Aldrich

Anti-CDCA2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PPP1R81, Anti-Repo-Man

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

VLSSKRRRISYQRDSDENLTDAEGKVIGLQIFNIDTDRACAVETSVDLSEISSKLGSTQSGFLVEESLPLSELTETSNALKVADCVVGKGSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDCA2(157313)

General description

Cell division cycle associated 2 (CDCA2) is expressed in the nucleus. The gene encoding it is localized on human chromosome 8p21.2.

Immunogen

Cell division cycle-associated protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Cell division cycle associated 2 (CDCA2) dephosphorylates histone H3 and modulates DNA damage responses. It has roles in nuclear envelope reformation and chromatin reorganization. The protein has been shown to be upregulated in neuroblastoma tumors and melanoma cell lines.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76428

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pauline Lagarde et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(5), 608-615 (2013-01-16)
Synovial sarcoma (SS) occurs in both children and adults, although metastatic events are much more common in adults. Whereas the importance of the t(X;18) translocation in SS oncogenesis is well established, the genetic basis of SS metastasis is still poorly
Paola Vagnarelli et al.
Nucleus (Austin, Tex.), 3(2), 138-142 (2012-05-05)
Coordination of late mitotic events is crucial for the maintenance of genome stability and for the control of gene expression after cell division. Reversible protein phosphorylation regulates this process by de-phosphorylation of mitotic phospho-proteins in a sequential and coordinated manner:
Paola Vagnarelli
Advances in experimental medicine and biology, 773, 401-414 (2014-02-25)
Nuclear structure and chromatin changes are very useful biomarkers in cancer diagnosis. Despite this, their biological significance and relevance to cancer progression are still not well understood. The identification of new proteins that link the nuclear envelope to chromatin organization
Fumihiko Uchida et al.
PloS one, 8(2), e56381-e56381 (2013-02-19)
Cell division cycle associated 2 (CDCA2) recruits protein phosphatase 1 to chromatin to antagonize activation of ataxia telangiectasia mutated (ATM)-dependent signal transduction. ATM kinase plays a critical role in the DNA damage response and its phosphorylation cascade to inhibit the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service