Skip to Content
Merck
All Photos(1)

Key Documents

B95-8

85011419, cotton-top tamarin monkey blood, Lymphoblast/Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

B95-8, from cotton-top tamarin monkey blood, 85011419, peripheral blood lymphocyte, Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)

biological source

cotton-top tamarin monkey blood

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast/Fibroblast

products

Epstein Barr Virus (EBV)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Cotton-top Tamarin Monkey peripheral blood lymphocyte

Cell Line Description

Derived from a cotton-top tamarin (Saguinus oedipus). Releases high titres of transforming Epstein Bar virus (EBV). Thus it provides a source of EBV to establish continuous lymphocytic lines from human donors. The cells should be handled under laboratory containment level 2.

In some instances the cell line B95-8 has been described as derived from ′marmoset′; however, this is not correct. B95-8 was derived from a cotton-top tamarin (Saguinas oedipus); this has been confirmed by DNA profiling.

Culture Medium

RPMI 1640 (R0883)+ 2 mM L-Glutamine (G7513) + 10% FBS / FCS (F2442).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml. May need to use 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. It is recommended that this cell line be frozen at no less than 8x1000,000 cells per ampoule.

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service