Skip to Content
Merck
All Photos(1)

Key Documents

AV39069

Sigma-Aldrich

Anti-NR2E3 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Nuclear receptor subfamily 2, group E, member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
human
citations:
2
technique(s):
western blot: suitable

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

45 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR2E3(10002)

Immunogen

Synthetic peptide directed towards the N terminal region of human NR2E3

Biochem/physiol Actions

NR2E3 is a member of nuclear receptor transcription factor family involved in embryonic development. It is a ligand-dependent retinal nuclear receptor and regulates the number of S-cones in the retina. Mutations in gene encoding NR2E3 cause S-cone syndrome and disrupt human cone photoreceptor mosaic.

Sequence

Synthetic peptide located within the following region: METRPTALMSSTVAAAAPAAGAASRKESPGRWGLGEDPTGVSPSLQCRVC

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sung Pyo Park et al.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie, 251(10), 2299-2309 (2013-04-23)
Enhanced S-cone syndrome is an orphan disease caused by mutations in the NR2E3 gene which result in an increased number of S-cones overpopulating the retina. Although the characteristic onset of enhanced S-cone syndrome can be well-documented by current ophthalmic imaging
Novel mutations in enhanced S-cone syndrome.
Kazuki Kuniyoshi et al.
Ophthalmology, 120(2), 431-431 (2013-02-05)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service