291S
MUC2 Positive Control Slides
suitable for immunohistochemistry (formalin-fixed, paraffin-embedded sections)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
packaging
pkg of 5 slides
Quality Level
manufacturer/tradename
Cell Marque™
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
shipped in
ambient
storage temp.
no temp limit
General description
MUC2 positive control slides are intended for use as positive controls for immunohistochemical (IHC) staining using an anti-MUC2 antibody.
Physical form
Paraffin embedded tissue section mounted on microscope slide
Other Notes
For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com
Control slide products are manufactured from human biological samples (HBS). Purchaser shall not make any attempt to trace or identify donors from whom the HBS material was obtained. Purchaser shall use any data (including genetic information), generated with or by use of the HBS only in compliance with obligations of confidentiality and non-use equivalent with all applicable international and national legislation and guidelines on the protection of personal data and confidentiality. In addition to the use restrictions contained on the product label and/or product datasheet or other applicable use terms and conditions of sale, Purchaser agrees to use the HBS solely in accordance with the World Medical Association (WMA) Declaration of Helsinki adopted in 1964 as subsequently amended by WMA on Ethical Principles for Medical Research Involving Human Subjects.
Control slide products are manufactured from human biological samples (HBS). Purchaser shall not make any attempt to trace or identify donors from whom the HBS material was obtained. Purchaser shall use any data (including genetic information), generated with or by use of the HBS only in compliance with obligations of confidentiality and non-use equivalent with all applicable international and national legislation and guidelines on the protection of personal data and confidentiality. In addition to the use restrictions contained on the product label and/or product datasheet or other applicable use terms and conditions of sale, Purchaser agrees to use the HBS solely in accordance with the World Medical Association (WMA) Declaration of Helsinki adopted in 1964 as subsequently amended by WMA on Ethical Principles for Medical Research Involving Human Subjects.
Legal Information
Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany
antibody
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service