Skip to Content
Merck
All Photos(1)

Key Documents

BCR5512

2,4-Dinitrophenylhydrazones, solutions, set

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

environmental

format

single component solution

storage temp.

−20°C

Packaging

set of 4x BCR-551 and 1x BCR-552 (blank)

Analysis Note

For more information please see:
BCR5512

Legal Information

BCR is a registered trademark of European Commission

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service