738484
Potassium 1-Boc-1H-indole-5-trifluoroborate
97%
About This Item
Assay
97%
form
solid
mp
190-195 °C
SMILES string
[K+].CC(C)(C)OC(=O)n1ccc2cc(ccc12)[B-](F)(F)F
InChI
1S/C13H14BF3NO2.K/c1-13(2,3)20-12(19)18-7-6-9-8-10(14(15,16)17)4-5-11(9)18;/h4-8H,1-3H3;/q-1;+1
InChI key
WQXAJDXDSOGMRW-UHFFFAOYSA-N
Application
- The cross-coupling reaction of 2(chloromethyl)-2,1-borazaronaphthalenes with trifluoroborates.
- Fluorination reactions of potassium salts of aryl and heteroaryl trifluoroborates.
- Nitrosation reactions of potassium trifluoroborates using nitrosonium tetrafluoroborate.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Skin Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service