Skip to Content
Merck
All Photos(1)

Key Documents

158461

Sigma-Aldrich

Dithiouracil

98%

Synonym(s):

2,4(1H,3H)-Pyrimidinedithione, 2,4-Dimercaptopyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2S2
CAS Number:
Molecular Weight:
144.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

279-281 °C (dec.) (lit.)

SMILES string

S=C1NC=CC(=S)N1

InChI

1S/C4H4N2S2/c7-3-1-2-5-4(8)6-3/h1-2H,(H2,5,6,7,8)

InChI key

ZEQIWKHCJWRNTH-UHFFFAOYSA-N

General description

Dithiouracil is a potential anticancer drug and its redox mechanism and electronic absorption behavior has been investigated in a wide pH range by UV-Vis spectroscopy, cyclic voltammetry and differential pulse voltammetry.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of 2-thiouracil and 2,4-dithiouracil for the determination of metal ions. Part I. Spectrophotometric determination of copper and silver.
G Misztal et al.
Annales Universitatis Mariae Curie-Sklodowska. Sectio D: Medicina, 38, 135-141 (1983-01-01)
Afzal Shah et al.
Journal of photochemistry and photobiology. B, Biology, 117, 269-277 (2012-11-06)
The redox mechanism and electronic absorption behavior of a commercial anticancer drug, 5-fluorouracil (5-FU) and two potential anticancer drugs, 2-thiouracil (2-TU) and dithiouracil (DTU) have been investigated in a wide pH range by UV-Vis spectroscopy, cyclic voltammetry and differential pulse
Marvin Pollum et al.
ChemMedChem, 13(10), 1044-1050 (2018-03-14)
Sulfur-substituted nucleobases (i.e., thiobases) are a prospective class of compounds for clinical and cosmetic topical phototherapies. Recent investigations of several thiobases have revealed the ultrafast and efficient population of reactive triplet states upon ultraviolet-A (UVA) irradiation and the subsequent generation
Wen-Jwu Wang et al.
Organic & biomolecular chemistry, 3(16), 3054-3058 (2005-09-28)
2,4-dithiouracil (DTU) forms in the crystals the H-bonded monohydrates of a 1:1:1 ratio with 18-crown-6 (18C6) 1, cis,syn,cis-isomer of dicyclohexano-18-crown-6 (DCH6A) 2, and benzo-18-crown-6 (B18C6) 3, while the anhydrous adduct with cis,anti,cis-isomer of dicyclohexano-18-crown-6 (DCH6B) 4 is of a 2:1
L Lapinski et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 54A(5), 685-693 (1998-07-29)
2,4-Pyrimidinedithiol (the dithiol form of 2,4-dithiouracil) was generated by UV (lambda > 335 nm) irradiation of the dithione form of 2,4-dithiouracil isolated in low-temperature argon or nitrogen matrices. The IR and UV spectra of the photoproduct are reported. The dithiol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service