Merck
  • KDM6B induces epithelial-mesenchymal transition and enhances clear cell renal cell carcinoma metastasis through the activation of SLUG.
KDM6B induces epithelial-mesenchymal transition and enhances clear cell renal cell carcinoma metastasis through the activation of SLUG.
International journal of clinical and experimental pathology (2015-08-12)
Qiang Li, Liejun Hou, Guofu Ding, Yinglong Li, Jiangping Wang, Biao Qian, Jianguo Sun, Qinzhang Wang
ABSTRACT
Clear cell renal cell carcinoma (ccRCC) is one of the most common kidney cancers; epithelial-mesenchymal transition (EMT) is associated with carcinoma invasion and metastasis. There have been several studies about the molecular regulation of EMT, but the relationship between histone demethylase and EMT is little known. Here, we reported KDM6B has high expression level in ccRCC and is positively correlated with poor ccRCC prognosis. KDM6B, also known as JMJD3, is a histone demethylase, can remove repressive histone H3K27me3 marks from chromatin, thereby activating gene expression. We found that the knockdown of KDM6B could inhibit ccRCC tumorigenesis in vitro; furthermore, KDM6B could induce EMT in ccRCC cells by activating the expression of master transcription factor SLUG. ChIP assays revealed that KDM6B stimulated SLUG expression by demethylate histone H3K27me3. The knockdown of KDM6B strongly inhibited ccRCC cell invasion in vitro, while the overexpression of KDM6B shown the opposite trend. Meanwhile, our analysis of the ccRCC tissue found that KDM6B expression was significantly corresponded with lymph node metastasis. Together, our data provide a novel epigenetic mechanism regulating tumor cell invasion and EMT, and provide a biomolecule for ccRCC diagnosis and prognosis.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
JMJD3/KDM6B Active human, recombinant, expressed in baculovirus infected insect cells, ≥70% (SDS-PAGE)