Key Documents
24843
Supelco
Calibrateur de débit pour Mini-Buck
model, M-5, flow rate 1-6000 mL/min
About This Item
Code UNSPSC :
41104008
Caractéristiques
model M-5
flow rate measuring range
1-6000 mL/min
Technique(s)
active air sampling: suitable
Application(s)
air monitoring
environmental
industrial hygiene
Description générale
Battery charger not included, order separately.
Application
Mini-buck flow calibrator is a gas flow calibrator, which may be used for the development and evaluation of a calibration gas generator based on the injection method for the analysis of volatile organic compounds in air.
À utiliser avec
Certificats d'analyse (COA)
Lot/Batch Number
Désolés, nous n'avons pas de COA pour ce produit disponible en ligne pour le moment.
Development and evaluation of a calibration gas generator for the analysis of volatile organic compounds in air based on the injection method.
Rosenberg E, et al.
Fresenius Journal of Analytical Chemistry, 371(6), 798-805 (2001)