Principaux documents
Z707406
ThinSeal film
non-sterile
About This Item
Code UNSPSC :
41122410
Nomenclature NACRES :
NB.22
Matériaux
polyester film
Stérilité
non-sterile
Conditionnement
pack of 100 ea
Fabricant/nom de marque
Excel Scientific 100-THIN-PLT
Technique(s)
ELISA: suitable
Longueur
141 mm
Larg. × L
7.94 cm × 14.1 cm
Largeur
79.4 mm
Description générale
Semi-pierceable, and leaves minimal residue. Polyester films just 25 μm thick, clearer and marginally pierceable with single-channel pipettes for sample recovery. Dimensions are 54.0 mm x 141.0 mm.
Informations légales
ThinSeal is a trademark of Excel Scientific, Inc.