Principaux documents
Z278432
Ace replacement cap
with hole
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Conditionnement
pk of 48 ea
Fabricant/nom de marque
Ace Glass 9590-46
Description générale
Caps with hole fit threaded outer 14/10 joints and, with an O-ring (Z504874), form an "O-RING-CAP SEAL" connection. Size: 20 mm.